Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552902 Supplier Invitrogen™ Supplier No. PA552902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHSAQDFYEN YEPKEILGRG VSSVVRRCIH KPTSQEYAVK VIDVTGGGSF SPEEVRELRE ATLKEVDILR KVSGHPNIIQ LKDTYETNTF FFLVFDLMKR GELFDYLTEK VTLSEKETRK IMR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

PHKG1 is a serine/threonine kinase with glycogenolytic regulatory functions. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. PHKG1 has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHKG1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHKG1
Gene Accession No. Q16816
Gene Alias gamma phosphorylase kinase; MGC137450 protein; PHGK; PHKG; PHKG1; PHKgammaM; PHK-gammaM; PHK-gamma-M; PHKgamma-M; PHKIN01; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase B kinase gamma catalytic chain, skeletal muscle isoform (Phosphorylase kinase gamma subunit 1); phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase; phosphorylase kinase catalytic subunit gamma 1; phosphorylase kinase gamma; phosphorylase kinase gamma 1; phosphorylase kinase gamma subunit; phosphorylase kinase gamma subunit 1; phosphorylase kinase subunit gamma 1; phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1; phosphorylase kinase, gamma 1 (muscle); serine/threonine-protein kinase PHKG1; unnamed protein product
Gene Symbols PHKG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHKG1. Recombinant protein control fragment (Product #RP-89806).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.