Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHKG1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986268
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6268 Supplier Bioss Supplier No. BS5009R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the Ser/Thr protein kinase family and encodes a protein with one protein kinase domain and two calmodulin-binding domains. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. This gene has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.

Specifications

Antigen PHKG1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PHKG1
Gene Accession No. Q16816
Gene Alias BOS_22899; gamma phosphorylase kinase; PHGK; Phkg; PHKG1; PHKgammaM; PHK-gammaM; PHK-gamma-M; PHKgamma-M; PHKIN01; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase B kinase gamma catalytic chain, skeletal muscle isoform (Phosphorylase kinase gamma subunit 1); Phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase catalytic subunit gamma 1; phosphorylase kinase gamma; phosphorylase kinase gamma 1; phosphorylase kinase gamma subunit; phosphorylase kinase gamma subunit 1; phosphorylase kinase subunit gamma 1; Phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1; phosphorylase kinase, gamma 1 (muscle); Serine/threonine-protein kinase PHKG1
Gene Symbols PHKG1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PHKG1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5260
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.