Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHKG2 Antibody (OTI1F9), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273373V
Description
PHKG2 Monoclonal specifically detects PHKG2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PHKG2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PHKG2 | |
Recombinant protein fragment corresponding to amino acids 1-216 of human PHKG2 (NP_000285) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1F9 | |
Western Blot, Immunohistochemistry | |
EC 2.7.11, EC 2.7.11.19, GSD9C, PHK-gamma-T, phosphorylase b kinase gamma catalytic chain, testis/liver isoform, Phosphorylase kinase subunit gamma-2, phosphorylase kinase, gamma 2 (testis), Phosphorylase kinase, gamma 2 (testis/liver), PSK-C3 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
5261.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction