Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHKG2 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA800354

Description
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. This isozyme may regulate glycogeneolysis in the testis.Specifications
PHKG2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P15735 | |
PHKG2 | |
Human recombit protein fragment corresponding to amino acids 1-216 of human PHKG2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2A3 | |
Unconjugated | |
PHKG2 | |
GSD9C | |
Mouse | |
Affinity Chromatography | |
RUO | |
5261 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction