Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596643 Supplier Invitrogen™ Supplier No. PA596643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQILMLRMIM EGQYQFSSPE WDDRSSTVKD LISRLLQVDP EARLTAEQAL QHPFFERCEG SQPWNLTPRQ RFRVAVWTVL AAGRVALSTH RVRPLTKNAL LRDPYALRSV RHLIDNCAFR LYGHWVKKGE QQNRAALFQH RPPGPFPIMG PEEEGDSAAI TEDEAVLVLG; Positive Samples: MCF7, 22RV1, HepG2, Jurkat, 293T, Mouse liver, Rat liver.

PHKG2 is a testis/liver isoform of the phosphorylase kinase gamma subunit. Phosphorylase kinase is a polymer of 16 subunits, four each of alpha, beta, gamma and delta. The alpha subunit includes the skeletal muscle and hepatic isoforms, encoded by two different genes. The beta subunit is the same in both the muscle and hepatic isoforms, and encoded by one gene. The gamma subunit also includes the skeletal muscle and hepatic isoforms, and the hepatic isoform is encoded by PHKG2. The delta subunit is a calmodulin and can be encoded by three different genes. The gamma subunits contain the active site of the enzyme, whereas the alpha and beta subunits have regulatory functions controlled by phosphorylation. The delta subunit mediates the dependence of the enzyme on calcium concentration. Mutations in PHKG2 cause glycogen storage disease type 9C, also known as autosomal liver glycogenosis. Alternatively spliced transcript variants encoding different isoforms have been identified in PHKG2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHKG2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHKG2
Gene Accession No. P15735, P31325, Q9DB30
Gene Alias 1500017I02Rik; GSD9C; MGC129049 protein; phkg2; PHK-gamma-LT; PHK-gamma-T; Phosphorylase b kinase gamma catalytic chain, liver/testis isoform; phosphorylase b kinase gamma catalytic chain, testis/liver isoform; phosphorylase kinase; phosphorylase kinase catalytic subunit gamma 2; phosphorylase kinase gamma subunit 2; phosphorylase kinase subunit gamma 2; phosphorylase kinase subunit gamma-2; phosphorylase kinase, gamma 2 (testis); Phosphorylase kinase, gamma 2 (testis/liver); PSK-C3; serine/threonine-protein kinase PHKG2; similar to phosphorylase kinase, gamma 2 (testis); zgc:55863; zgc:55863 protein
Gene Symbols PHKG2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 237-406 of human PHKG2 (NP_0002851).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140671, 5261, 68961
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.