Learn More
Invitrogen™ PHLDA1 Monoclonal Antibody (OTI4D9)

Description
PHLDA1 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PHLDA1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4D9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PHLDA1 |
| Gene Accession No. | Q8WV24 |
| Gene Alias | apoptosis-associated nuclear protein; DT1P1B11; PHLDA1; PHRIP; pleckstrin homology like domain family A member 1; pleckstrin homology-like domain family A member 1; pleckstrin homology-like domain, family A, member 1; PQR protein; PQ-rich protein; proline- and glutamine-rich protein; Proline- and histidine-rich protein; proline-histidine rich protein; T-cell death associated; T-cell death-associated gene 51 protein; Tdag; TDAG51 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.