Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHLDA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100750 Supplier Invitrogen™ Supplier No. PA5100750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PHLDA1.

Cytotoxic T lymphocyte (CTL)-mediated cytotoxicity constitutes an important component of specific effector mechanisms in immunosurveillance against virus-infected or -transformed cells. Two mechanisms appear to account for this activity, one of which is the perforin-based process. Independently, a FAS-based mechanism involves the transducing molecule FAS (APO-1) and its ligand (FAS-L). The human FAS (APO-1) protein is a 48 kDa cell surface glycoprotein that belongs to a family of receptors that includes CD40, nerve growth factor receptors and tumor necrosis factor receptors. The FAS antigen is expressed on a broad range of lymphoid cell lines, and is expressed at high levels in T cells subsequent to crosslinking of the T cell receptor (TCR). A previously undescribed protein, TDAG51, restores activation- induced apoptosis in cells that have lost the ability to display Fas in response to activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHLDA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PHLDA1
Gene Accession No. Q62392, Q8WV24, Q9QZA1
Gene Alias apoptosis-associated nuclear protein; DT1P1B11; PHLDA1; PHRIP; pleckstrin homology like domain family A member 1; pleckstrin homology-like domain family A member 1; pleckstrin homology-like domain, family A, member 1; PQR protein; PQ-rich protein; proline- and glutamine-rich protein; Proline- and histidine-rich protein; proline-histidine rich protein; T-cell death associated; T-cell death-associated gene 51 protein; Tdag; TDAG51
Gene Symbols PHLDA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PHLDA1(Accession Q8WV24), corresponding to amino acid residues M1-G45.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21664, 22822, 29380
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.