Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHLDA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592845 Supplier Invitrogen™ Supplier No. PA592845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKSPDEVLRE GELEKRSDSL FQLWKKKRGV LTSDRLSLFP ASPRARPKEL RFHSILKVDC VERTGKYVYF TIVTTDHKEI DFRCAGESCW NAAIALALID FQNRRALQDF RSRQERTAPA APAEDAVAAA AAAPSEPSEP SRPSPQPKPR TP; Positive Samples: SW620, SKOV3, HeLa; Cellular Location: Cytoplasm, Membrane, Peripheral membrane protein.

TSSC3 (Tumor-suppressing STF cDNA 3 protein), also known as PHLDA2 (pleckstrin homology-like domain family A member 2), BWR1C, HLDA2 or IPL, is a cytoplasmic protein that is involved in fetal and placental growth. Expressed at high levels in placenta and adult prostate and at low levels in liver, lung and brain, TSSC3 is an apoptosis-related protein that acts as a negative growth regulator and is expressed during normal human development. TSSC3, a protein expressed from the maternal allele, is imprinted on placenta, liver and fetal tissues during embryogenesis and is removed once development is complete. Defects or alterations in the gene encoding TSSC3 are associated with several afflictions such as lung, ovarian and breast cancer, rhabdomyosarcoma, Beckwith-Wiedemann syndrome, Wilms tumor, low birth weight and adrenocortical carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHLDA2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHLDA2
Gene Accession No. Q53GA4
Gene Alias Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene C protein; BRW 1C; BRW1C; BWR 1C; BWR1C; HLDA 2; HLDA2; Imprinted in placenta and liver protein; IPL; p17-Beckwith-Wiedemann region 1 C; p17-Beckwith-Wiedemann region 1C; p17-BWR1C; PHLDA2; pleckstrin homology like domain family A member 2; pleckstrin homology-like domain family A member 2; pleckstrin homology-like domain, family A, member 2; Pphlda2; protein 50C15; TSSC 3; TSSC3; tumor suppressing subchromosomal transferable fragment cDNA 3; tumor suppressing subtransferable candidate 3; Tumor-suppressing STF cDNA 3 protein; tumor-suppressing subchromosomal transferable fragment 3; tumor-suppressing subchromosomal transferable fragment candidate gene 3 protein; tumor-supressing STF cDNA 3
Gene Symbols PHLDA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-152 of human PHLDA2 (NP_0033021).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7262
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.