Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHLDA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100751
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100751 Supplier Invitrogen™ Supplier No. PA5100751
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PHLDA3.

The exact function of PHLDA3 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHLDA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PHLDA3
Gene Accession No. Q5PQT7, Q9WV95, Q9Y5J5
Gene Alias Phlda3; pleckstrin homology like domain family A member 3; pleckstrin homology-like domain family A member 3; pleckstrin homology-like domain, family A, member 2; pleckstrin homology-like domain, family A, member 3; TDAG/lpl homolog 1; TDAG51/Ipl homolog 1; TIH1
Gene Symbols PHLDA3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PHLDA3(Accession Q9Y5J5), corresponding to amino acid residues N95-S127.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23612, 27280, 363989
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.