Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHLDB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111530 Supplier Invitrogen™ Supplier No. PA5111530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTLTLYPCGN ACTIDGLPVR QPTRLTQGCM LCLGQSTFLR FNHPAEAKWM KSMIPAGGRA PGPPYSPVPA ESESLVNGNH T Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 94%.

PHLDB1 (Pleckstrin Homology Like Domain Family B Member 1) is a Protein Coding gene expressed in the pituitary gland. Diseases associated with PHLDB1 include Mixed Glioma and Li-Fraumeni Syndrome. An important paralog of this gene is PHLDB2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHLDB1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHLDB1
Gene Accession No. Q86UU1
Gene Alias AU041016; D330037A14Rik; DLNB07; KIAA0638; Ll5; LL5A; Ll5alpha; Phldb1; pleckstrin homology like domain family B member 1; pleckstrin homology-like domain family B member 1; pleckstrin homology-like domain family B member 1 variant 3; pleckstrin homology-like domain family B member 1 variant 4; pleckstrin homology-like domain, family B, member 1; protein LL5-alpha
Gene Symbols PHLDB1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human PHLDB1. Recombinant protein control fragment (Product #RP-107072).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.