Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHLDB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544287
Description
Peptide sequence: GPSGFPPLMH HSILHHLPAG RERGEEGEHA YDTLSLESSD SMETSISTGG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
PHLDB1 (Pleckstrin Homology Like Domain Family B Member 1) is a Protein Coding gene expressed in the pituitary gland. Diseases associated with PHLDB1 include Mixed Glioma and Li-Fraumeni Syndrome. An important paralog of this gene is PHLDB2.
Specifications
PHLDB1 | |
Polyclonal | |
Unconjugated | |
PHLDB1 | |
AU041016; D330037A14Rik; DLNB07; KIAA0638; Ll5; LL5A; Ll5alpha; Phldb1; pleckstrin homology like domain family B member 1; pleckstrin homology-like domain family B member 1; pleckstrin homology-like domain family B member 1 variant 3; pleckstrin homology-like domain family B member 1 variant 4; pleckstrin homology-like domain, family B, member 1; protein LL5-alpha | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23187 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86UU1 | |
PHLDB1 | |
Synthetic peptide directed towards the middle region of human PHLDB1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction