Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloroglucinol, 99+%, anhydrous

Catalog No. p-4510299
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 108-73-6 | C6H6O3 | 126.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-73-6
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.1
InChI Key QCDYQQDYXPDABM-UHFFFAOYSA-N
Synonym phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxy
PubChem CID 359
ChEBI CHEBI:16204
IUPAC Name benzene-1,3,5-triol
SMILES C1=C(C=C(C=C1O)O)O

Specifications

Melting Point 215.0°C to 218.0°C
Color White to Beige
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula (OH)3C6H3
Quantity 25 g
Fieser 01,47; 06,147
Merck Index 15,7439
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone ethanol, ether and pyridine
Formula Weight 126.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 203-611-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.