Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phloroglucinol, anhydrous, 98%

Click to view available options
Quantity:
50g
250g
108-73-6
C6H6O3
126.111
MFCD00002286
QCDYQQDYXPDABM-UHFFFAOYSA-N
phloroglucinol,1,3,5-benzenetriol,1,3,5-trihydroxybenzene,phloroglucin,phloroglucine,spasfon-lyoc,s-trihydroxybenzene,benzene-s-triol,5-hydroxyresorcinol,benzene, trihydroxy
359
CHEBI:16204
benzene-1,3,5-triol
C1=C(C=C(C=C1O)O)O
108-73-6
C6H6O3
126.111
MFCD00002286
QCDYQQDYXPDABM-UHFFFAOYSA-N
phloroglucinol,1,3,5-benzenetriol,1,3,5-trihydroxybenzene,phloroglucin,phloroglucine,spasfon-lyoc,s-trihydroxybenzene,benzene-s-triol,5-hydroxyresorcinol,benzene, trihydroxy
359
CHEBI:16204
benzene-1,3,5-triol
C1=C(C=C(C=C1O)O)O
Applications
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .

Solubility
Soluble in diethyl ether, ethanol and pyridine.

Notes
Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
Melting Point 215°C to 220°C
Quantity 250g
Beilstein 1341907
Merck Index 14,7328
Formula Weight 126.11
Percent Purity 98%
Chemical Name or Material Phloroglucinol, anhydrous

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SY1050000

TSCA : TSCA