Learn More
Phloroglucinol, anhydrous, 98%
Description
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .
Solubility
Soluble in diethyl ether, ethanol and pyridine.
Notes
Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
Specifications
Specifications
| Melting Point | 215°C to 220°C |
| Quantity | 50 g |
| Beilstein | 1341907 |
| Sensitivity | Hygroscopic; Light sensitive |
| Merck Index | 14,7328 |
| Solubility Information | Soluble in diethyl ether,ethanol and pyridine. |
| Formula Weight | 126.11 |
| Percent Purity | 98% |
| Chemical Name or Material | Phloroglucinol, anhydrous |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 203-611-2 |
| RTECSNumber | SY1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.