Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloroglucinol dihydrate, 98%

Catalog No. p-7020918
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 6099-90-7 | C6H10O5 | 162.141 g/mol

Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6099-90-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00149090
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol;dihydrate
SMILES C1=C(C=C(C=C1O)O)O.O.O

Specifications

Melting Point 214°C to 217°C
Odor Odorless
Quantity 25 g
Sensitivity Light sensitive
Merck Index 14,7328
Solubility Information Slightly soluble in water.
Formula Weight 162.14 (126.11 Anhydrous)
Percent Purity 98%
Chemical Name or Material Phloroglucinol dihydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-611-2
RTECSNumber SY1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.