Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloroglucinol Dihydrate, 99%

Catalog No. p-3737218
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 6099-90-7 | C6H6O3·2H2O | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6099-90-7
Molecular Formula C6H6O3·2H2O
Molecular Weight (g/mol) 162.14
MDL Number MFCD00149090
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol;dihydrate
SMILES C1=C(C=C(C=C1O)O)O.O.O

Specifications

Melting Point 218.0°C to 221.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H3(OH)3·2H2O
Quantity 25 g
Beilstein 06, 1092
Merck Index 15, 7439
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and pyridine
Formula Weight 162.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.