Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloxine B, Certified, 85%, Thermo Scientific™

Catalog No. ac417870250
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 18472-87-2
Molecular Formula C20 H2 Br4 Cl4 Na2 O5
MDL Number MFCD00005061
Synonym Acid Red 92, C.I. 45410

Specifications

Absorption Ratio 1.38 to 2.24
Dye Content 80% min.
Assay Percent Range 85%
Beilstein 19, IV, 2921
Merck Index 15, 7440
Infrared Spectrum Authentic
Lambda Max 546 to 550nm
Formula Weight 829.64
Grade Pure
Packaging Glass bottle
Color Brown/Red to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Phloxine B
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.