Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHLPP Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281766C
This item is not returnable.
View return policy
Description
PHLPP Polyclonal antibody specifically detects PHLPP in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
PHLPP | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
DyLight 650 | |
EC 3.1.3.16, hSCOP, KIAA0606, MGC161555, PH domain and leucine rich repeat protein phosphatase, PH domain and leucine rich repeat protein phosphatase 1, PH domain leucine-rich repeat-containing protein phosphatase 1, PHLPP, pleckstrin homology domain containing, family E (with leucine rich repeats)member 1, Pleckstrin homology domain-containing family E member 1, PLEKHE1, SCN circadian oscillatory protein, SCOPPH domain-containing family E member 1, Suprachiasmatic nucleus circadian oscillatory protein | |
PHLPP antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PHLPP. The immunogen is located within amino acids 250 - 300 of PHLPP. Amino Acid Squence: REPAEPPPEAGPRLAPPE | |
0.1 mL | |
Cancer, Protein Phosphatase, Tumor Suppressors | |
23239 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction