Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHLPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103165 Supplier Invitrogen™ Supplier No. PA5103165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCOP.

PHLPP1 is a member of the serine/threonine phosphatase family, which are important regulators of Akt serine-threonine kinases (AKT1, AKT2, AKT3) and conventional/novel protein kinase C (PKC) isoforms. PHLPP1 and PHLPP2 have a similar domain structure and have been shown to dephosphorylate and inactivate, distinct Akt isoforms, at one of the two critical phosphorylation sites required for activation: Serine473. PHLPP2 dephosphorylates AKT1 and AKT3, whereas PHLPP1 is specific for AKT2 and AKT3. PHLPP1 promotes apoptosis and may act as a tumor suppressor. Increased expression of PHLPP1 may also play a role in obesity and type 2 diabetes by interfering with Akt-mediated insulin signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHLPP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PHLPP1
Gene Accession No. O60346, Q8CHE4, Q9WTR8
Gene Alias AI836256; Circadian Oscillatory Protein (SCOP); EC 3.1.3.16; hSCOP; KIAA0606; mKIAA0606; PH domain and leucine rich repeat protein phosphatase 1; PH domain leucine-rich repeat protein phosphatase 1; PH domain leucine-rich repeat-containing protein phosphatase 1; PH domain-containing family E member 1; Phlpp; PHLPP1; pleckstrin homology domain containing, family E (with leucine rich repeats) member 1; pleckstrin homology domain-containing family E member 1; pleckstrin homology domain-containing family E protein 1; Plekhe1; PPM3A; protein phosphatase, Mg2+/Mn2+ dependent 3A; SCN circadian oscillatory protein; SCOP; Suprachiasmatic nucleus circadian oscillatory protein
Gene Symbols PHLPP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PHLPP1(Accession O60346), corresponding to amino acid residues G1438-S1488.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23239, 59265, 98432
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.