Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHLPP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$330.00 - $547.00
Specifications
Antigen | PHLPP2 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PHLPP2 Polyclonal antibody specifically detects PHLPP2 in Human samples. It is validated for ImmunofluorescenceSpecifications
PHLPP2 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Tumor Suppressors | |
PBS, pH 7.2, 40% glycerol | |
23035 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 3.1.3.16, KIAA0931PH domain leucine-rich repeat-containing protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase-like, PH domain leucine-rich repeat-containing protein phosphatase-like, PHLPPLPHLPP-like | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DLHLVLCTVETPASEICAGEGRESLYLQLHGDLVRRLEPTERPLQIVYDYLSRLGFDDPVRIQEEATNPDLGCMIRFYG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title