Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHLPP2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518758
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Protein phosphatase involved in regulation of Akt and PKC signaling. Mediates dephosphorylation in the C-terminal domain hydrophobic motif of members of the AGC Ser/Thr protein kinase family; specifically acts on 'Ser-473' of AKT1, 'Ser-660' of PRKCB isoform beta-II and 'Ser-657' of PRKCA. Akt regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes. Dephosphorylation of 'Ser-473' of Akt triggers apoptosis and decreases cell proliferation. Also controls the phosphorylation of AKT3. Dephosphorylates STK4 on 'Thr-387' leading to STK4 activation and apoptosis (PubMed:20513427). Dephosphorylates RPS6KB1 and is involved in regulation of cap-dependent translation (PubMed:21986499). Inhibits cancer cell proliferation and may act as a tumor suppressor. Dephosphorylation of PRKCA and PRKCB leads to their destabilization and degradation. Dephosphorylates RAF1 inhibiting its kinase activity (PubMed:24530606). [UniProt].
Specifications
| PHLPP2 | |
| Polyclonal | |
| Unconjugated | |
| Phlpp2 | |
| AI481772; C130044A18Rik; GTPase activating protein and VPS9 domains 1; KIAA0931; PH domain and leucine rich repeat protein phosphatase 2; PH domain leucine-rich repeat-containing protein phosphatase 2; PH domain leucine-rich repeat-containing protein phosphatase-like; PHLPP2; Phlppl; PHLPP-like; PPM3B; protein phosphatase, Mg2+/Mn2+ dependent 3B; RGD1562857 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 23035 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6ZVD8 | |
| Phlpp2 | |
| Synthetic peptide sequence (PHEEDRTEPPEEFD) corresponding to the C-terminus amino acids of PHLPPL (aa 1307-1321). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction