Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHLPP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111278
Description
Immunogen sequence: DLHLVLCTVE TPASEICAGE GRESLYLQLH GDLVRRLEPT ERPLQIVYDY LSRLGFDDPV RIQEEATNPD LGCMIRFYG Highest antigen sequence indentity to the following orthologs: Rat - 41%, Mouse - 96%.
Protein phosphatase involved in regulation of Akt and PKC signaling. Mediates dephosphorylation in the C-terminal domain hydrophobic motif of members of the AGC Ser/Thr protein kinase family; specifically acts on 'Ser-473' of AKT1, 'Ser-660' of PRKCB isoform beta-II and 'Ser-657' of PRKCA. Akt regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes. Dephosphorylation of 'Ser-473' of Akt triggers apoptosis and decreases cell proliferation. Also controls the phosphorylation of AKT3. Dephosphorylates STK4 on 'Thr-387' leading to STK4 activation and apoptosis (PubMed:20513427). Dephosphorylates RPS6KB1 and is involved in regulation of cap-dependent translation (PubMed:21986499). Inhibits cancer cell proliferation and may act as a tumor suppressor. Dephosphorylation of PRKCA and PRKCB leads to their destabilization and degradation. Dephosphorylates RAF1 inhibiting its kinase activity (PubMed:24530606). [UniProt].
Specifications
| PHLPP2 | |
| Polyclonal | |
| Unconjugated | |
| Phlpp2 | |
| AI481772; C130044A18Rik; GTPase activating protein and VPS9 domains 1; KIAA0931; PH domain and leucine rich repeat protein phosphatase 2; PH domain leucine-rich repeat-containing protein phosphatase 2; PH domain leucine-rich repeat-containing protein phosphatase-like; PHLPP2; Phlppl; PHLPP-like; PPM3B; protein phosphatase, Mg2+/Mn2+ dependent 3B; RGD1562857 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZVD8 | |
| Phlpp2 | |
| Recombinant protein fragment of Human PHLPP2 (Product #RP-98598) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction