Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHLPPL Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558152
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-155-8152 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8152 Supplier Bethyl Laboratories, Inc Supplier No. A300661AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

PHLPP2 is a protein phosphatase that specifically mediates dephosphorylation of ′Ser-473′ of AKT1. This protein regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes. Dephosphorylation of ′Ser-473′ of AKT1 triggers apoptosis and decrease cell proliferation. It also controls the phosphorylation of AKT3.

Specifications

Antigen PHLPPL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PHLPP2
Gene Accession No. Q6ZVD8, Q8BXA7
Gene Alias PH domain leucine-rich repeat protein phosphatase-like; PHLPP-like; PHLPP2; PHLPPL; KIAA0931
Gene Symbols Phlpp2
Host Species Rabbit
Immunogen Between 1275 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23035, 244650
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.