Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHLPPL Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody

Supplier:  Bethyl Laboratories, Inc A300661A

Encompass_Preferred

Catalog No. 50-155-8150


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

PHLPP2 is a protein phosphatase that specifically mediates dephosphorylation of ′Ser-473′ of AKT1. This protein regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes. Dephosphorylation of ′Ser-473′ of AKT1 triggers apoptosis and decrease cell proliferation. It also controls the phosphorylation of AKT3.
Specifications

Specifications

PHLPPL
Polyclonal
Unconjugated
PHLPP2
PH domain leucine-rich repeat protein phosphatase-like; PHLPP-like; PHLPP2; PHLPPL; KIAA0931
Rabbit
Antigen affinity chromatography
RUO
23035, 244650
4° C
Liquid
Immunoprecipitation, Western Blot
0.2 mg/ml
TBS with 0.1% BSA and 0.09% sodium azide
Q6ZVD8, Q8BXA7
Phlpp2
Between 1275 and C-term
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.