Learn More
Invitrogen™ pho Polyclonal Antibody

Description
pho Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | pho |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | pho |
| Gene Accession No. | Q8ST83 |
| Gene Alias | Polycomb protein PHO pho CG17743 |
| Gene Symbols | pho |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.