Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phoenix Diagnostics Olympus™ AU Series ISE Reagents
Shop All Phoenix Diagnostics, Inc Products
Click to view available options
Type:
Buffer
Cleaning Solution
High Serum Standard
Internal Reference Solution
Low Serum Standard
Low/High Urine Solution
Mid Standard
NA+/K+ Check Solution
Reference
Wash Solution
Description
- For the quantitative determination of sodium, potassium and chloride concentrations in serum and urine samples
- Solutions available include buffer, mid-standard, reference, low serum standard, high serum standard, low/high urine standard, internal reference fill, Na/K selectivity check, cleaning and wash
Specifications
Specifications
| Type | Cleaning Solution |
| For Use With (Equipment) | Olympus™ AU series analyzer |
| Capacity (Metric) | 4 x 100 mL |
| Reagents | Cleaning Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction