Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Phorbol 12,13-dibutyrate
SDP

Catalog No. RD10195093 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10195093 10 mg
41-531 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RD10195093 Supplier R&D Systems™ Supplier No. 4153/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Tocris product, part of R&D Systems. Protein kinase C activator

Phorbol 12,13-dibutyrate is a protein kinase C activator. Induces contraction of vascular smooth muscle and inhibits MLC phosphatase (MLCP) in vascular smooth muscle. Activity does not alter intracellular Ca2+ concentration. Also inhibits the activity of Na+,K+ ATPase in OK cells.

For more information about how Phorbol 12,13-dibutyrate may be used, see our protocol:

Generation of β cells from hPSCs

Chemical Identifiers

CAS 37558-16-0
Molecular Formula C28H40O8

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 504.61
Purity >98%
Molecular Weight (g/mol) 504.62
Chemical Name or Material Phorbol 12, 13-dibutyrate
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.