Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phorbol 12-myristate 13-acetate, 97%

Catalog No. p-3737215
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 16561-29-8 | C36H56O8 | 616.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16561-29-8
Molecular Formula C36H56O8
Molecular Weight (g/mol) 616.84
MDL Number MFCD00036736
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
PubChem CID 27924
ChEBI CHEBI:37537
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O

Specifications

Melting Point 72.0°C
Color White to Yellow
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 mg
Merck Index 14,7332
Formula Weight 616.84
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phorbol 12-myristate 13-acetate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.