Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phorbol 12-myristate 13-acetate, MP Biomedicals™

Catalog No. MP218388201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218388201 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP218388201 Supplier MP Biomedicals, Inc Supplier No. 0218388201
Add to Cart
Add to Cart

Used to increase the biosynthesis of Heme and the repair and creation of iron-sulfur clusters by delivering Fe(2+) to proteins involved in these pathways

  • PMA (Phorbol 12-myristate 13-acetate) activates protein kinase C (PKC) and is a known TPA (tumor promoting agent)
  • PMA has shown chromosome-breaking (clastogenic) activity
  • Phorbol 12-myristate 13-acetate (PMA) has been shown to induce the activity of PLD1 (phospholipase D1)
  • PMA is a tumor promoter, as seen with protein kinase C (PKC) signaling pathway associated with: gene transcription, cell growth, cell differentiation, programmed cell death, immune pathway, receptor desensitization
  • Phorbol 12-myristate 13-acetate is used to study: growth factors, hormones, cytokines-mediated regulation of cellular growth and differentiation
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Melting Point 50°C to 70°C
Color White
Purity ≥98%
Molecular Weight (g/mol) 616.8
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.