Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phos-ERK1 (T202/Y204)/ERK2 (T185/Y187) Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC7806G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7806G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7806G Supplier R&D Systems Supplier No. IC7806G
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ERK1/ERK2 Monoclonal specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen ERK1/ERK2 (p Tyr187, p Tyr204, p Thr185, p Thr202)
Applications Flow Cytometry
Classification Monoclonal
Clone 269434
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Rabbit
Immunogen Phosphopeptide containing the ERK1 T202/Y204 site
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human, mouse, and rat ERK1 and ERK2 dually phosphorylated at T202/Y204 or T185/Y187, respectively.
Target Species Human, Mouse, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.