Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffered Saline, pH 7.6, Ricca Chemical
SDP

Catalog No. 58217625 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5821-762-5 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5821-762-5 Supplier Ricca Chemical Company Supplier No. 5821.762.5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10049-21-5 , 1310-73-2 , 67-56-1 , 75-05-8 , 7732-18-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Color Colorless
Grade Laboratory
Concentration or Composition (by Analyte or Components)
  • Acetonitrile: 27.02%
  • Methyl Alcohol: 26.99%
  • Sodium Phosphate, Monobasic Monohydrate: 0.22%
Packaging Cubitainer
pH 7.6
Quantity 10 L
Physical Form Liquid
Chemical Name or Material Phosphate / Acetonitrile / Methanol Solution
ShelfLife 12 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.