Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer Solution, pH 6.8, for Phenols, Ricca Chemical
SDP

Catalog No. p-7239389
Click to view available options
Quantity:
500 mL
1 L

For the Determination of Phenols

Specifications

Chemical Name or Material Phosphate Buffer Solution
CAS 10049-21-5
Concentration 1X
For Use With (Application) Water Analysis
pH 7
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.