Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphate Buffer Solution, pH 7, ASTM for Chlorine, Ricca Chemical

Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Phosphate Buffer Solution |
Name Note | pH 7 |
CAS | 7732-18-5, 7558-79-4, 7778-77-0, 1310-73-2 |
CAS Min % | 92.0, 3.14, 2.33, 0.34 |
Color | Colorless |
CAS Max % | 96.0, 3.27, 2.43, 0.36 |
Concentration | 1X |
For Use With (Application) | Water Analysis |
pH | 7 |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction