Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
GSA_VA

Catalog No. p-27643 Shop All Fisherbrand Products
Click to view available options
Quantity:
500mL
1L
4L
20L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly Cube
This item is not returnable. View return policy
This item is not returnable. View return policy

Commonly used buffer

  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.119M phosphates
  • pH of a 1X solution made from this concentrated solution is ∼7.4. To prepare a 1X solution, mix one volume of Fisher BioReagents 10X PBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH
  • Intended for laboratory research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Concentration 10 X
Physical Form Liquid
Quantity 20L
Solution Type Phosphate Buffered Saline
Packaging Poly Cube
Grade Biochemical
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.