Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Phosphate-buffered Saline (PBS, 10X), pH 7.4 PROMO
Click to view available options
:
4 L
Specifications
Specifications
Chemical Name or Material | Phosphate Buffered Saline (PBS) |
Color | Colorless |
Concentration | 10 X |
pH | 7.4 |
Physical Form | Liquid |
Quantity | 4 L |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction