Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), RNase free

Catalog No. p-7082332
Click to view available options
Quantity:
250 mL
500 mL

Catalog No. AAJ62851AP

Supplier: Thermo Scientific Chemicals J62851AP

Only null left
Add to Cart

Helps to maintain a constant pH

Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. Phosphate-buffered saline with EDTA is used to disengage attached and clumped cells. It is also used in cell culturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
Physical Form Liquid
Quantity 500 mL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.