Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 5X), with EDTA
Description
PBS with EDTA is also used to disengage attached and clumped cells. PBS-EDTA is used as a buffer in procedures generating dendritic cells from monocytes. PBS+EDTA to get better cell sorting. PBS/EDTA may also be used directly without addition of trypsine for very gentle cell dissociation. PBS can be used to take a reference spectrum when measuring the protein adsorption in ellipsometry. PBS can be used as a diluent in methods to dry biomolecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Chemical Name or Material | Phosphate Buffered Saline (PBS) |
Concentration | 5 X |
Physical Form | Liquid |
Quantity | 1 L |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.