Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Phosphate Kit
SDPSCI_ED

Catalog No. s98239 Shop All LaMotte Company Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S98239 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Phosphate reagents are considered to be potential health hazards.
Catalog No. S98239 Supplier LaMotte™ Supplier No. 312102
Only null left
Add to Cart
Add to Cart
Phosphate reagents are considered to be potential health hazards.

Use in freshwater or salt water.

  • Low range phosphate test, 0.2 to 2.0 ppm
  • Ascorbic acid reagent system produces a blue color in the presence of phosphate
  • Enough reagent for 50 tests

Specifications

For Use With (Application) Use for freshwater and salt water
Includes Ascorbic acid octet comparator with axial reader
Class Size 1 Class
Product Type LaMotte Phosphate Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.