Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Standard, 500 ppm PO43- (163 ppm P), Ricca Chemical
SDP

Catalog No. p-2916791 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Amber Glass Bottle
Equivalence 1mL = 0.5mg PO4,500ppm PO4,163ppm Phosphorus (P)
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Phosphate Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.