Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Standard, 10,000 ppm PO43- (3261 ppm P), Ricca Chemical
SDP

Catalog No. r5839500100 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL

Chemical Identifiers

CAS 7778-77-0, 7732-18-5
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Packaging Natural Poly Bottle
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-component Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Phosphate Standard, 1mL = 10mg PO43-,10,000ppm PO43- (3,261ppm P)
Show More Show Less
Recommended Storage 15°C - 30°C (59°F - 86°F)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.