Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Standard, 1000 ppm PO43- (326 ppm P), Ricca Chemical
SDP

Catalog No. 583916 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Natural Poly Bottle
Equivalence 1mL = 1mg PO4,1000ppm PO4,326.1ppm Phosphorus (P)
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Phosphate Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.