Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphatidylcholine, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
Description
Spectrum™ Chemical Phosphatidylcholine , also known as lecithin, has been shown to play a vital role in many important areas including maintaining cell structure, fat metabolism, memory, nerve signaling, as a precursor to important neurotransmitters, and liver health.
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 1 kg |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction