Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phosphatidylinositol 3,4,5-trisphosphate Monoclonal Antibody (RC6F8)
GREENER_CHOICE

Catalog No. A21328
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21328 Supplier Invitrogen™ Supplier No. A21328
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Phosphatidylinositol 3,4,5-trisphosphate Monoclonal Antibody for ICC/IF

Phosphatidylinositol (3, 4, 5)-trisphosphate (PtdIns(3, 4, 5)P3) commonly abbreviated to PIP3 is the product of the class I phosphoinositide 3-kinases (PI 3-kinases) activity on phosphatidylinositol (4, 5)-bisphosphate. PtdIns (3, 4, 5) P3 is dephophosphorylated by the phosphatase, PTEN on the 3 position and by SHIPs (SH2-containing inositol phosphatase) on the 5' position of the inositol ring. The PH domain in a number of proteins binds to PtdIns (3, 4, 5) P3. Such proteins include Akt/PKB, PDK1, Btk1 and ARNO. The generation of PtdIns (3, 4, 5) P3 at the plasma membrane upon the activation of class I PI 3-kinases causes these proteins to translocate to the plasma membrane and accordingly affects their activity. The PH domain allows binding between PtdIns (3, 4, 5) P3 and G-protein coupled receptor kinases (GRKs). This enhances the binding of the GRK to the plasma membrane. [National Center for Biotechnology Information. PubChem Compound Database; CID=53477782].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphatidylinositol 3,4,5-trisphosphate
Applications Immunocytochemistry
Classification Monoclonal
Clone RC6F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 5mM sodium azide; pH 7.2
Gene Alias C12H24O22P4; phosphatidylinositol 3; phosphatidylinositol-3; PIP3
Host Species Mouse
Immunogen phosphatidylinositol 3,4,5-diphosphate.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Target Molecule Chemical
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.