Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phosphatidylinositol 4,5-biphosphate Monoclonal Antibody
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA126438
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosphatidylinositol 4,5-bisphosphate or PIP2 is an important second messenger for a number of signaling processes. When certain extracellular signals are received, Phosphilipase C cleaves phosphatidylinositol into DAG and PIP2. PIP2 has been shown to interact with actin structural proteins including profiling, gelsolin, and cofilin, and demonstrates regulatory function of actin polymerization. It has also been shown to regulate vesicular transport by interacting with proteins like ADP-ribosylation factor and phospholipase D. PIP2 has also been shown to significantly affect intracellular calcium concentrations, which also regulate a variety of activities.
Specifications
Phosphatidylinositol 4,5-biphosphate | |
Monoclonal | |
Unconjugated | |
C45H85O19P3; PIP2 | |
Phosphatidylinositol-4,5-biphosphate (PIP2) from bovine spinal cord. | |
50 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |
ELISA, Radioimmune Assays (RIA) | |
1 mg/mL | |
PBS with 10% BSA and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction