Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-14-3-3 (S58) Antibody, R&D Systems™

Catalog No. PPS042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS042 Supplier R&D Systems Supplier No. PPS042
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide coupled columns.

14-3-3 Polyclonal specifically detects 14-3-3 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Sheep, Xenopus, Zebrafish samples. It is validated for Western Blot.

Specifications

Antigen 14-3-3 (p ser58)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias 1433, 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Gene Symbols YWHAQ
Host Species Rabbit
Immunogen Phosphopeptide corresponding to amino acid residues surrounding the phospho-S58 of 14-3-3
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10971
Test Specificity This antibody is specific for the 29 kDa 14-3-3 phosphorylated at S58 in Western blots of rat brain stem lysates.
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Sheep, Xenopus species, Zebrafish
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.