Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-5-Lipoxygenase (Ser271) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985250
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5250 Supplier Bioss Supplier No. BS3251R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.

Specifications

Antigen Phospho-5-Lipoxygenase (Ser271)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ALOX5
Gene Accession No. P09917
Gene Alias 5 - Lipoxygenase; 5-lipoxygenase; 5LO; 5-LO; 5-LOX; 5LPG; 5LX; AI850497; ALOX5; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; BOS_24317; F730011J02; leukotriene A4 synthase; LOC100341714; LOG5; LOX-5; LOX5A; RP11-67C2.3
Gene Symbols ALOX5
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from rat 5-Lipoxygenase around the phosphorylation site of Ser271.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.