Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-53BP2 (Ser698) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105070 Supplier Invitrogen™ Supplier No. PA5105070
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of ASPP2 only when phosphorylated at Ser698.

Regulator that plays a central role in regulation of apoptosis and cell growth via its interactions. Regulates TP53 by enhancing the DNA binding and transactivation function of TP53 on the promoters of proapoptotic genes in vivo. Inhibits the ability of APPBP1 to conjugate NEDD8 to CUL1, and thereby decreases APPBP1 ability to induce apoptosis. Impedes cell cycle progression at G2/M. Its apoptosis-stimulating activity is inhibited by its interaction with DDX42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-53BP2 (Ser698)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TP53BP2
Gene Accession No. Q13625
Gene Alias 53BP2; Apoptosis-stimulating of p53 protein 2; apoptosis-stimulating protein of p53, 2; ASPP2; BBP; Bcl2-binding protein; P PPP1R13A; P53BP2; PPP1R13A; Renal carcinoma antigen NY-REN-51; TP53BP2; transformation related protein 53 binding protein 2; Trp53bp2; tumor protein p53 binding protein 2; tumor protein p53 binding protein, 2; tumor suppressor p53-binding protein 2
Gene Symbols TP53BP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TP53BP2(Accession Q13625), corresponding to amino acid residues around phosphorylated Ser698.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305025, 7159
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.