Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-A-Raf/B-Raf/C-Raf (Ser446, Ser338, Ser299) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143756 Supplier Invitrogen™ Supplier No. PA5143756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to phospho-B-Raf (Ser-579) peptide before affinity purification using phospho-B-Raf (Ser-446) peptide. The purified antibody detects a strong band at 95 kDa* and a weaker band at 105 kDa in western blots of human Jurkat or A431 cells treated with Calyculin A. The antibody also detects other phosphorylated proteins at 150 and 200 kDa in calyculin A treated cells.

The Ras-Raf-MAP kinase signaling pathway is involved in control of cell proliferation and differentiation. The Raf kinase family includes A-Raf, B-Raf, and C-Raf. Each family member has three highly conserved regions (CR1-3). The N-terminal CR1 contains the Ras-GTP-binding domain. The CR2 contains a negative regulatory serine residue (C-Raf (Ser259)/B-Raf(Ser365)) that may bind 14-3-3 proteins. The CR3 is the catalytic domain that contains phosphorylation sites for Raf-regulating enzymes within two segments, the N-region and the activation segment. Activation of C-Raf involves phosphorylation at many sites including Ser-338, Tyr-341, and multiple catalytic domain sites. In B-Raf, multiple phosphorylation sites have been identified, but their specific roles are uncertain. Phosphorylation of Ser-446 may prime B-Raf for activation, and Ser-446 and/or Ser-447 phosphorylation may be critical for B-Raf biological activity during PC12 differentiation. Ser-579 is required for growth factor activation and kinase activity. Thus, multiple sites of phosphorylation within Rafs may be important for regulation of their activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-A-Raf/B-Raf/C-Raf (Ser446, Ser338, Ser299)
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene BRAF
Gene Accession No. P04049, P11345, P15056, P28028, Q99N57
Gene Alias BRAF1; CRaf; Raf1; RAFB1
Gene Symbols BRAF, RAF1
Host Species Rabbit
Immunogen Phospho-B-Raf (Ser-446) synthetic peptide (coupled to KLH) corresponds to amino acids surrounding serine 446 in human B-Raf. This sequence is conserved in B-Rafs from many species, and has high homology to C-Raf (Ser-338) and A-Raf (Ser-299).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109880, 110157, 114486, 24703, 5894, 673
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.