Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-A-Raf/B-Raf/C-Raf (Ser471, Ser579, Ser432) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143757 Supplier Invitrogen™ Supplier No. PA5143757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to phospho-C-Raf (Ser-338) peptide before affinity purification using phospho-C-Raf (Ser-471) peptide. The purified antibody detects a band at 74 kDa* corresponding to C-Raf in western blots of human Jurkat cells treated with Calyculin A, but is not observed after lambda phosphatase treatment.

The Ras-Raf-MAP kinase signaling pathway is involved in control of cell proliferation and differentiation. The Raf kinase family includes A-Raf, B-Raf, and C-Raf. Each family member has three highly conserved regions (CR1-3). The N-terminal CR1 contains the Ras-GTP-binding domain. The CR2 contains a negative regulatory serine residue (C-Raf (Ser259)/B-Raf(Ser365)) that may bind 14-3-3 proteins. The CR3 is the catalytic domain that contains phosphorylation sites for Raf-regulating enzymes within two segments, the N-region and the activation segment. Activation of C-Raf involves phosphorylation at many sites including Ser-338, Tyr-341, and multiple catalytic domain sites. In B-Raf, multiple phosphorylation sites have been identified, but their specific roles are uncertain. Phosphorylation of Ser-446 may prime B-Raf for activation, and Ser-446 and/or Ser-447 phosphorylation may be critical for B-Raf biological activity during PC12 differentiation. Ser-579 is required for growth factor activation and kinase activity. Thus, multiple sites of phosphorylation within Rafs may be important for regulation of their activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-A-Raf/B-Raf/C-Raf (Ser471, Ser579, Ser432)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene BRAF
Gene Accession No. P04049, P11345, P15056, P28028, Q99N57
Gene Alias BRAF1; CRaf; Raf1; RAFB1
Gene Symbols BRAF, RAF1
Host Species Rabbit
Immunogen A synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Ser-471 in human C-Raf. This sequence has high homology with similar regions in rat and mouse C-Raf, and has high homology to the conserved sites in B-Raf (Ser-579) and A-Raf (Ser-432).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109880, 110157, 114486, 24703, 5894, 673
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.