Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-AANAT (Thr29) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1761
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1761 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1761 Supplier Invitrogen™ Supplier No. PA1761
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-761 detects phospho serotonin N-acetyltransferase (AA-NAT) from rat tissues. PA1-761 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~23 kDa protein representing phospho AA-NAT in rat pineal gland whole protein extract. PA1-761 immunizing phosphopeptide (Cat. # PEP-064) corresponds to amino-terminal amino acid residues 24-33 of mouse AA-NAT. Total anti-amino-terminal AA-NAT-specific IgG (Cat. # PA1-760) was affinity purified on resin-coupled non-phosphopeptide (Cat. # PEP-063). Phospho-specific amino-terminal IgG (Cat. # PA1-761) was subsequently isolated on resin-coupled phosphopeptide (Cat. # PEP-064). This sequence is completely conserved between mouse, rat, and human AA-NAT protein.

Arylalkylamine N-Acetyltransferase (AANAT) is the penultimate enzyme is melatonin synthesis. Hormonal melatonin has been implicated in sleep and circadian clock function. Large changes in the abundance of AANAT are believed to be responsible for the large day/night rhythms in melatonin synthesis in the pineal gland and retina. Most research shows that AANAT activity is regulated by controlling the steady-state levels of the protein. Recent research, however, suggests that another mechanism may also play a role in controlling melatonin production without altering AANAT protein and activity. This mechanism may explain the sudden physiological increase in circulating melatonin observed at the onset of the dark period in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-AANAT (Thr29)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Aanat
Gene Accession No. Q64666
Gene Alias Aanat; AA-NAT; Aralkylamine N-acetyltransferase; arylakylamine N-acetyltransferase; Arylalkylamine N - acetyltransferase (Serotonin N - acetyltransferase); arylalkylamine N-acetyltransferase; DSPS; Nat-2; Nat4; Seretonin N-acetyltransferase; serotonin acetylase; Serotonin N-acetyltransferase; SNAT
Gene Symbols Aanat
Host Species Rabbit
Immunogen Synthetic Peptide: C(24) Q R R H T(p) L P A S(33).
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25120
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.